Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162999.2 | NP_001156471.1 | 15 - 16 | 3198 | 94 | |
GenBank mRNA | AK052684.1 | - | 1 - 2 | 648 | 94 | |
AK122512.1 | - | 7 - 8 | 2275 | 94 | ||
BC116316.1 | - | 8 - 9 | 2462 | 94 | ||
BC116317.1 | - | 8 - 9 | 2462 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |