Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025794.2 | NP_080070.2 | 5 - 6 | 814 | 74 | |
GenBank mRNA | AK002483.1 | - | 5 - 6 | 767 | 74 | |
AK037969.1 | - | 5 - 6 | 731 | 74 | ||
AK075673.1 | - | 5 - 6 | 765 | 74 | ||
AK141684.1 | - | 5 - 6 | 768 | 74 | ||
AK152624.1 | - | 5 - 6 | 783 | 74 | ||
AK169686.1 | - | 5 - 6 | 719 | 74 | ||
BC012522.1 | - | 5 - 6 | 763 | 2648103 | 74 | |
BC057670.1 | - | 5 - 6 | 781 | 2810976 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |