Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctso | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177662.2 | NP_808330.1 | 1 - 2 | 105 | ||
GenBank mRNA | AK034490.1 | - | 1 - 2 | 105 | ||
AK040071.1 | - | 1 - 2 | 105 | |||
AK049470.1 | - | 1 - 2 | 105 | |||
AK143683.1 | - | 1 - 2 | 105 | |||
AK165930.1 | - | 1 - 2 | 105 | |||
AK166103.1 | - | 1 - 2 | 105 | |||
BC044664.1 | - | 1 - 2 | 105 | |||
BC098219.1 | - | 1 - 2 | 1066724 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |