Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpini2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026460.3 | NP_080736.2 | 6 - 7 | 1017 | 105 | |
GenBank mRNA | AF251276.1 | - | 6 - 7 | 1002 | 105 | |
AK007347.1 | - | 6 - 7 | 1018 | 105 | ||
AK007510.1 | - | 6 - 7 | 992 | 105 | ||
BC098466.1 | - | 6 - 7 | 994 | 6433250 | 105 | |
CF581490.1 | - | 3 - 4 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |