Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172515.2 | NP_766103.2 | 1 - 2 | 137 | 141 | |
GenBank mRNA | AK029899.1 | - | 1 - 2 | 146 | 141 | |
AK133102.1 | - | 1 - 2 | 188 | 141 | ||
BC050789.1 | - | 1 - 2 | 137 | 6706914 | 141 | |
BC100435.1 | - | 1 - 2 | 137 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |