Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133701.2 | NP_598462.1 | 10 - 11 | 1449 | 92 | |
GenBank mRNA | AK081998.1 | - | 10 - 11 | 1384 | 92 | |
AK145461.1 | - | 10 - 11 | 1452 | 92 | ||
AK166890.1 | - | 10 - 11 | 1401 | 92 | ||
BC014869.1 | - | 10 - 11 | 1376 | 3602158 | 92 | |
BC023691.1 | - | 10 - 11 | 1378 | 5344332 | 92 | |
BC025030.1 | - | 10 - 11 | 1395 | 4947416 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |