Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027656.2 | NP_081932.1 | 3 - 4 | 1434 | 70 | |
GenBank mRNA | AK016700.1 | - | 3 - 4 | 799 | 70 | |
AK038975.1 | - | 2 - 3 | 315 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons |