Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198169.2 | NP_937812.1 | 2 - 3 | 250 | 78 | |
GenBank mRNA | AB221583.1 | - | 1 - 2 | 130 | 78 | |
AB221584.1 | - | 1 - 2 | 130 | 78 | ||
BC029763.1 | - | 1 - 2 | 73 | 78 | ||
BC059066.1 | - | 2 - 3 | 250 | 6825428 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |