Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153680.2 | NP_710147.1 | 12 - 13 | 1275 | 94 | |
GenBank mRNA | AK077650.1 | - | 12 - 13 | 1277 | 94 | |
AK077780.1 | - | 12 - 13 | 1277 | 94 | ||
AK156299.1 | - | 12 - 13 | 1326 | 94 | ||
BC023732.1 | - | 12 - 13 | 1291 | 5352355 | 94 | |
BC023925.1 | - | 7 - 8 | 738 | 94 | ||
BC026571.1 | - | 12 - 13 | 1275 | 4986268 | 94 | |
BC029675.1 | - | 12 - 13 | 1262 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |