Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Preb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016703.3 | NP_057912.2 | 7 - 8 | 1223 | 59 | |
GenBank mRNA | AF150808.1 | - | 7 - 8 | 1126 | 59 | |
AF193017.1 | - | 7 - 8 | 1135 | 59 | ||
AK029838.1 | - | 7 - 8 | 1159 | 59 | ||
AK036866.1 | - | 7 - 8 | 1161 | 59 | ||
AK151288.1 | - | 7 - 8 | 1205 | 59 | ||
AK206719.1 | - | 2 - 3 | 179 | 59 | ||
BC017527.1 | - | 7 - 8 | 1126 | 4921119 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |