Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133918.2 | NP_598679.1 | 5 - 6 | 2648 | 131 | |
GenBank mRNA | AK029337.1 | - | 5 - 6 | 2619 | 131 | |
AK052368.1 | - | 5 - 6 | 2728 | 131 | ||
AK088995.1 | - | 5 - 6 | 2714 | 132 | ||
AK140388.1 | - | 3 - 4 | 2064 | 131 | ||
AK155486.1 | - | 5 - 6 | 2950 | 131 | ||
BC005481.1 | - | 5 - 6 | 2648 | 3485057 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |