Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153135.3 | NP_694775.1 | 16 - 17 | 2791 | 87 | |
GenBank mRNA | AJ487854.1 | - | 16 - 17 | 2792 | 87 | |
AK220417.1 | - | 6 - 7 | 1132 | 87 | ||
BC137615.1 | - | 16 - 17 | 2582 | 8860634 | 87 | |
BC144783.1 | - | 14 - 15 | 2339 | 9053331 | 87 | |
BC144784.1 | - | 14 - 15 | 2360 | 9053332 | 87 | |
BC145695.1 | - | 13 - 14 | 2366 | 40130998 | 87 | |
FJ824810.1 | - | 15 - 16 | 2670 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |