Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023663.6 | NP_076152.2 | 5 - 6 | 875 | 80 | |
GenBank mRNA | AF302127.3 | - | 5 - 6 | 878 | 80 | |
AK077233.1 | - | 5 - 6 | 1059 | 80 | ||
AK145203.1 | - | 5 - 6 | 857 | 80 | ||
BC057871.1 | - | 5 - 6 | 838 | 5294197 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |