Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008467.4 | NP_032493.1 | 1 - 2 | 357 | 156 | |
GenBank mRNA | AF020771.1 | - | 1 - 2 | 65 | 156 | |
AK147920.1 | - | 1 - 2 | 366 | 156 | ||
BC026821.1 | - | 1 - 2 | 152 | 3672708 | 156 | |
BC052162.1 | - | 1 - 2 | 358 | 6510795 | 156 | |
CT010281.1 | - | 1 - 2 | 65 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |