Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290425.1 | NP_001277354.1 | 1 - 2 | 53 | 113 | |
NM_152825.2 | NP_690038.1 | 1 - 2 | 53 | 113 | ||
GenBank mRNA | AK040660.1 | - | 1 - 2 | 54 | 113 | |
AK162825.1 | - | 1 - 2 | 54 | 113 | ||
BB213271.2 | - | 1 - 2 | 65 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mus musculus 1700 Probe spans exons |