Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172808.2 | NP_766396.2 | 12 - 13 | 1256 | 90 | |
GenBank mRNA | AK077206.1 | - | 12 - 13 | 1257 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |