Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159963.1 | NP_001153435.1 | 2 - 3 | 400 | 98 | |
NM_178729.4 | NP_848844.1 | 2 - 3 | 400 | 98 | ||
GenBank mRNA | AK029957.1 | - | 2 - 3 | 348 | 98 | |
AK041497.1 | - | 2 - 3 | 372 | 98 | ||
AK054009.1 | - | 2 - 3 | 370 | 98 | ||
AK082931.1 | - | 2 - 3 | 370 | 98 | ||
AK085100.1 | - | 2 - 3 | 373 | 98 | ||
AK088067.1 | - | 2 - 3 | 367 | 98 | ||
AK150664.1 | - | 2 - 3 | 360 | 98 | ||
AK159744.1 | - | 2 - 3 | 389 | 98 | ||
AK171361.1 | - | 2 - 3 | 354 | 98 | ||
AK171783.1 | - | 2 - 3 | 260 | 98 | ||
BC047214.1 | - | 313 | 6466511 | 98 | ||
BC113798.1 | - | 2 - 3 | 318 | 40062506 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |