Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007646.4 | NP_031672.2 | 2 - 3 | 420 | 76 | |
GenBank mRNA | AK038439.1 | - | 2 - 3 | 398 | 76 | |
AK040498.1 | - | 2 - 3 | 406 | 76 | ||
AK042970.1 | - | 2 - 3 | 389 | 76 | ||
AK150412.1 | - | 2 - 3 | 438 | 76 | ||
AK151016.1 | - | 2 - 3 | 432 | 76 | ||
AK151918.1 | - | 2 - 3 | 389 | 76 | ||
AK153215.1 | - | 2 - 3 | 436 | 76 | ||
AK153334.1 | - | 2 - 3 | 389 | 76 | ||
BC046312.1 | - | 2 - 3 | 396 | 6310453 | 76 | |
CT010386.1 | - | 2 - 3 | 371 | 76 | ||
L11332.1 | - | 2 - 3 | 377 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |