Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290383.1 | NP_001277312.1 | 5 - 6 | 931 | 144 | |
NM_194055.3 | NP_918944.2 | 5 - 6 | 931 | 147 | ||
GenBank mRNA | AK042164.1 | - | 5 - 6 | 543 | 147 | |
AK140848.1 | - | 5 - 6 | 932 | 144 | ||
BC031468.1 | - | 3 - 4 | 234 | 4037840 | 147 | |
BC059280.1 | - | 3 - 4 | 232 | 147 | ||
DV659240.1 | - | 3 - 4 | 287 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |