Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293667.1 | NP_001280596.1 | 16 - 17 | 2009 | 74 | |
NM_001293668.1 | NP_001280597.1 | 16 - 17 | 2009 | 74 | ||
NM_015804.3 | NP_056619.1 | 16 - 17 | 2009 | 74 | ||
NR_121603.1 | 16 - 17 | 2009 | 74 | |||
GenBank mRNA | AF156551.1 | - | 16 - 17 | 1801 | 74 | |
AK147497.1 | - | 16 - 17 | 2012 | 74 | ||
AK173088.1 | - | 16 - 17 | 2162 | 74 | ||
BC036322.1 | - | 1 - 2 | 70 | 74 | ||
BC138715.1 | - | 16 - 17 | 1728 | 8861737 | 74 | |
BC145399.1 | - | 16 - 17 | 1729 | 74 | ||
BC145400.1 | - | 16 - 17 | 1729 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |