Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198031.1 | NP_932148.1 | 5 - 6 | 611 | 88 | |
GenBank mRNA | AK051775.1 | - | 5 - 6 | 623 | 88 | |
AK150149.1 | - | 5 - 6 | 635 | 88 | ||
AK165503.1 | - | 5 - 6 | 634 | 88 | ||
AK171491.1 | - | 5 - 6 | 617 | 88 | ||
AK171804.1 | - | 5 - 6 | 626 | 88 | ||
BC046615.1 | - | 5 - 6 | 611 | 88 | ||
BC058566.1 | - | 5 - 6 | 611 | 6466057 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |