Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008407.2 | NP_032433.2 | 1 - 2 | 129 | 86 | |
GenBank mRNA | AK146700.1 | - | 1 - 2 | 130 | 86 | |
BC015276.1 | - | 1 - 2 | 4212290 | 86 | ||
X70393.1 | - | 1 - 2 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |