Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081221.1 | NP_001074690.1 | 14 - 15 | 2785 | 68 | |
GenBank mRNA | BC132447.1 | - | 10 - 11 | 1724 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |