Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031373.3 | NP_113550.3 | 5 - 6 | 510 | 62 | |
GenBank mRNA | AF303894.1 | - | 5 - 6 | 458 | 62 | |
AK008230.1 | - | 5 - 6 | 475 | 62 | ||
AK149146.1 | - | 5 - 6 | 457 | 62 | ||
BC003875.1 | - | 5 - 6 | 441 | 3495186 | 62 | |
BC049965.1 | - | 5 - 6 | 456 | 3155091 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |