Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183161.3 | NP_898984.3 | 7 - 8 | 878 | 89 | |
GenBank mRNA | CF165170.1 | - | 4 - 5 | 364 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Detect Genomic DNA Mus musculus Probe spans exons |