Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159774.1 | NP_001153246.1 | 18 - 19 | 1969 | 81 | |
NM_010546.2 | NP_034676.1 | 18 - 19 | 1969 | 81 | ||
GenBank mRNA | AF026524.1 | - | 17 - 18 | 1837 | 81 | |
AF088910.1 | - | 18 - 19 | 2029 | 81 | ||
AK144603.1 | - | 18 - 19 | 1962 | 81 | ||
AK151955.1 | - | 18 - 19 | 1921 | 81 | ||
AK153151.1 | - | 18 - 19 | 1921 | 81 | ||
AK155587.1 | - | 16 - 17 | 4320 | 81 | ||
AK156294.1 | - | 18 - 19 | 1922 | 81 | ||
AK219506.1 | - | 3 - 4 | 232 | 81 | ||
BC004772.1 | - | 18 - 19 | 1907 | 81 | ||
BC037723.1 | - | 18 - 19 | 1941 | 4482634 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |