Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027338.1 | NP_081614.1 | 10 - 11 | 848 | 130 | |
GenBank mRNA | AK150754.1 | - | 10 - 11 | 869 | 130 | |
AK151035.1 | - | 10 - 11 | 869 | 130 | ||
AK161865.1 | - | 900 | 130 | |||
BC010811.1 | - | 10 - 11 | 848 | 4210911 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |