Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027338.1 | NP_081614.1 | 1 - 2 | 110 | 71 | |
GenBank mRNA | AK008946.1 | - | 1 - 2 | 148 | 71 | |
AK150754.1 | - | 1 - 2 | 131 | 71 | ||
AK151035.1 | - | 1 - 2 | 131 | 71 | ||
AK156241.1 | - | 124 | 71 | |||
AK161865.1 | - | 162 | 71 | |||
BC010811.1 | - | 1 - 2 | 110 | 4210911 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |