Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027338.1 | NP_081614.1 | 3 - 4 | 246 | 56 | |
GenBank mRNA | AK008946.1 | - | 3 - 4 | 284 | 56 | |
AK150754.1 | - | 3 - 4 | 267 | 56 | ||
AK151035.1 | - | 3 - 4 | 267 | 56 | ||
AK156241.1 | - | 260 | 56 | |||
AK161865.1 | - | 298 | 56 | |||
BC010811.1 | - | 3 - 4 | 246 | 4210911 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |