Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027338.1 | NP_081614.1 | 4 - 5 | 364 | 105 | |
GenBank mRNA | AK008946.1 | - | 4 - 5 | 402 | 105 | |
AK150754.1 | - | 4 - 5 | 385 | 105 | ||
AK151035.1 | - | 4 - 5 | 385 | 105 | ||
AK156241.1 | - | 378 | 105 | |||
AK161865.1 | - | 416 | 105 | |||
BC010811.1 | - | 4 - 5 | 364 | 4210911 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |