Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010612.2 | NP_034742.2 | 10 - 11 | 1680 | 81 | |
GenBank mRNA | AK141938.1 | - | 10 - 11 | 1703 | 81 | |
BC020530.1 | - | 10 - 11 | 1680 | 4238984 | 81 | |
EU884114.1 | - | 10 - 11 | 1710 | 81 | ||
S53103.1 | - | 10 - 11 | 1691 | 81 | ||
X59397.1 | - | 10 - 11 | 1627 | 81 | ||
X70842.1 | - | 10 - 11 | 1705 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |