Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010612.2 | NP_034742.2 | 7 - 8 | 1244 | 93 | |
GenBank mRNA | AK054510.1 | - | 7 - 8 | 1277 | 93 | |
AK141938.1 | - | 7 - 8 | 1267 | 93 | ||
BC020530.1 | - | 7 - 8 | 1244 | 4238984 | 93 | |
EU884114.1 | - | 7 - 8 | 1274 | 93 | ||
S53103.1 | - | 7 - 8 | 1255 | 93 | ||
X59397.1 | - | 7 - 8 | 1191 | 93 | ||
X70842.1 | - | 7 - 8 | 1269 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |