Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clock | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289826.1 | NP_001276755.1 | 12 - 13 | 1223 | 146 | |
NM_007715.6 | NP_031741.1 | 12 - 13 | 1266 | 146 | ||
GenBank mRNA | AB221635.1 | - | 9 - 10 | 878 | 146 | |
AF000998.1 | - | 12 - 13 | 1266 | 146 | ||
AK030352.1 | - | 12 - 13 | 1166 | 146 | ||
AK129118.1 | - | 12 - 13 | 1223 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |