Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289770.1 | NP_001276699.1 | 1 - 2 | 148 | 83 | |
NM_001289771.1 | NP_001276700.1 | 1 - 2 | 148 | 83 | ||
NM_027270.2 | NP_081546.1 | 1 - 2 | 148 | 83 | ||
GenBank mRNA | AK042620.1 | - | 1 - 2 | 102 | 83 | |
AK155980.1 | - | 1 - 2 | 102 | 83 | ||
BC064811.1 | - | 1 - 2 | 136 | 5686167 | 83 | |
BC087543.1 | - | 1 - 2 | 84 | 6334680 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |