Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199122.1 | NP_001186051.1 | 3 - 4 | 449 | 69 | |
NM_078478.5 | NP_510963.1 | 3 - 4 | 402 | 69 | ||
GenBank mRNA | AF412297.1 | - | 3 - 4 | 346 | 69 | |
AK005636.1 | - | 3 - 4 | 346 | 69 | ||
AK093887.1 | - | 3 - 4 | 378 | 69 | ||
AK134592.1 | - | 3 - 4 | 332 | 69 | ||
AK146433.1 | - | 3 - 4 | 309 | 69 | ||
AK158904.1 | - | 3 - 4 | 392 | 69 | ||
AK159695.1 | - | 3 - 4 | 376 | 69 | ||
AK159724.1 | - | 3 - 4 | 376 | 69 | ||
AK164122.1 | - | 3 - 4 | 347 | 69 | ||
AK167970.1 | - | 3 - 4 | 373 | 69 | ||
AK168022.1 | - | 3 - 4 | 379 | 69 | ||
AK168678.1 | - | 3 - 4 | 450 | 69 | ||
BC008622.1 | - | 3 - 4 | 380 | 3588758 | 69 | |
BC010224.1 | - | 3 - 4 | 382 | 3500685 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |