Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020295.3 | NP_064691.2 | 5 - 6 | 619 | 100 | |
GenBank mRNA | AF190665.1 | - | 5 - 6 | 421 | 100 | |
AK030631.1 | - | 6 - 7 | 1425 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |