Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153598.2 | NP_705826.1 | 2 - 3 | 893 | 148 | |
GenBank mRNA | AK145951.1 | - | 2 - 3 | 903 | 148 | |
BC028826.1 | - | 2 - 3 | 893 | 5098890 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |