Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500005072.mm.1 |
TC0500000547.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005072.mm.1 |
NM_001081103.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005072.mm.1 |
NM_001081103.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005074.mm.1 |
TC0500000547.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005074.mm.1 |
NM_001081103.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005074.mm.1 |
NM_001081103.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005076.mm.1 |
TC0500000547.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005076.mm.1 |
NM_001081103.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005076.mm.1 |
NM_001081103.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005071.mm.1 |
TC0500000547.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005071.mm.1 |
NM_001081103.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005071.mm.1 |
NM_001081103.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005073.mm.1 |
TC0500000547.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005073.mm.1 |
NM_001081103.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005073.mm.1 |
NM_001081103.2 |
Refseq |