Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shcbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011369.2 | NP_035499.1 | 9 - 10 | 1359 | 141 | |
GenBank mRNA | AF017152.1 | - | 9 - 10 | 1359 | 141 | |
AK135226.1 | - | 1 - 2 | 1981 | 141 | ||
AK137496.1 | - | 8 - 9 | 1841 | 141 | ||
AK144966.1 | - | 9 - 10 | 1382 | 141 | ||
AK162943.1 | - | 9 - 10 | 1373 | 141 | ||
BC070455.1 | - | 9 - 10 | 1379 | 30606785 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |