Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195496.1 | NP_001182425.1 | 13 - 14 | 2537 | 97 | |
NM_001195497.1 | NP_001182426.1 | 12 - 13 | 2489 | 97 | ||
NM_001195498.1 | NP_001182427.1 | 13 - 14 | 2540 | 97 | ||
NM_001195499.1 | NP_001182428.1 | 13 - 14 | 2537 | 97 | ||
NM_027539.5 | NP_081815.3 | 12 - 13 | 2492 | 97 | ||
GenBank mRNA | AB198721.1 | - | 13 - 14 | 1894 | 97 | |
AK049179.1 | - | 12 - 13 | 1941 | 97 | ||
AK082633.1 | - | 12 - 13 | 1988 | 97 | ||
AY968049.1 | - | 13 - 14 | 1825 | 97 | ||
AY968050.1 | - | 13 - 14 | 1828 | 97 | ||
AY968051.1 | - | 13 - 14 | 1825 | 97 | ||
BC052071.1 | - | 5 - 6 | 533 | 97 | ||
BC056921.1 | - | 12 - 13 | 2460 | 6822681 | 97 | |
BC060192.1 | - | 12 - 13 | 1793 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |