Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199467.2 | NP_955761.2 | 12 - 13 | 1963 | 128 | |
GenBank mRNA | AK089531.1 | - | 5 - 6 | 703 | 128 | |
AK144799.1 | - | 11 - 12 | 1762 | 129 | ||
AK166072.1 | - | 12 - 13 | 1974 | 128 | ||
AY512923.1 | - | 7 - 8 | 1097 | 128 | ||
BC051674.1 | - | 3 - 4 | 410 | 128 | ||
BC094429.1 | - | 11 - 12 | 1307 | 128 | ||
BC137910.1 | - | 7 - 8 | 873 | 8860929 | 128 | |
BC145028.1 | - | 7 - 8 | 873 | 9053576 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |