Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030733.3 | NP_109658.2 | 1 - 2 | 258 | 88 | |
GenBank mRNA | AK036257.1 | - | 1 - 2 | 259 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mus musculus Probe spans exons |