Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026194.4 | NP_080470.2 | 1 - 2 | 148 | 91 | |
GenBank mRNA | AK007993.2 | - | 1 - 2 | 93 | 91 | |
AK032663.1 | - | 1 - 2 | 84 | 91 | ||
AK076206.1 | - | 1 - 2 | 93 | 91 | ||
AK129211.1 | - | 1 - 2 | 90 | 91 | ||
AK132697.1 | - | 1 - 2 | 112 | 91 | ||
BC058809.1 | - | 1 - 2 | 87 | 91 | ||
BC080203.1 | - | 1 - 2 | 126 | 91 | ||
BC099529.1 | - | 1 - 2 | 136 | 91 | ||
BC132195.1 | - | 1 - 2 | 100 | 40130472 | 91 | |
BC138153.1 | - | 1 - 2 | 100 | 8861172 | 91 | |
BY097965.1 | - | 1 - 2 | 149 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |