Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011299.4 | NP_035429.1 | 13 - 14 | 1437 | 86 | |
GenBank mRNA | AJ131021.1 | - | 13 - 14 | 1265 | 86 | |
AK019881.1 | - | 13 - 14 | 1342 | 86 | ||
AK050996.1 | - | 5 - 6 | 829 | 86 | ||
BC038251.1 | - | 7 - 8 | 664 | 86 | ||
BC043064.1 | - | 13 - 14 | 1387 | 5706153 | 86 | |
BC051079.1 | - | 13 - 14 | 1347 | 6491001 | 86 | |
BC055331.1 | - | 13 - 14 | 1437 | 6406879 | 86 | |
BC056946.1 | - | 13 - 14 | 1380 | 6822831 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |