Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177909.5 | NP_808577.3 | 1 - 2 | 339 | 73 | |
GenBank mRNA | AK037058.1 | - | 1 - 2 | 323 | 73 | |
AK156494.1 | - | 1 - 2 | 300 | 73 | ||
AK172687.1 | - | 1 - 2 | 311 | 73 | ||
BC099942.1 | - | 1 - 2 | 313 | 30103308 | 73 | |
BC153825.1 | - | 1 - 2 | 317 | 40093280 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |