Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199018.2 | NP_950183.1 | 4 - 5 | 433 | 85 | |
GenBank mRNA | AK149830.1 | - | 4 - 5 | 364 | 85 | |
AK150259.1 | - | 4 - 5 | 85 | |||
BC062944.1 | - | 4 - 5 | 348 | 30357177 | 85 | |
BC078461.1 | - | 3 - 4 | 198 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |