Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207633.2 | NP_997516.1 | 2 - 3 | 630 | 74 | |
GenBank mRNA | AK020780.1 | - | 2 - 3 | 204 | 74 | |
AK030473.1 | - | 2 - 3 | 632 | 74 | ||
AK045453.1 | - | 2 - 3 | 342 | 74 | ||
AK053562.1 | - | 2 - 3 | 631 | 74 | ||
AK079683.1 | - | 2 - 3 | 242 | 74 | ||
AK143152.1 | - | 2 - 3 | 200 | 74 | ||
AK160251.1 | - | 3 - 4 | 258 | 74 | ||
BC048712.1 | - | 2 - 3 | 265 | 6328657 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |