Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133747.2 | NP_598508.2 | 4 - 5 | 667 | 96 | |
GenBank mRNA | AK012997.1 | - | 4 - 5 | 601 | 96 | |
AK046481.1 | - | 4 - 5 | 584 | 96 | ||
AK146215.1 | - | 4 - 5 | 611 | 96 | ||
AK146761.1 | - | 4 - 5 | 633 | 96 | ||
AK146955.1 | - | 4 - 5 | 580 | 96 | ||
AK167224.1 | - | 4 - 5 | 551 | 96 | ||
BC004626.1 | - | 4 - 5 | 564 | 3585698 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |