Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008777.3 | NP_032803.2 | 5 - 6 | 732 | 75 | |
GenBank mRNA | AK145839.1 | - | 5 - 6 | 597 | 75 | |
AK149519.1 | - | 5 - 6 | 598 | 75 | ||
BC013458.1 | - | 5 - 6 | 590 | 4223155 | 75 | |
X51942.1 | - | 5 - 6 | 557 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |