Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028315.2 | NP_082591.2 | 17 - 18 | 2432 | 120 | |
GenBank mRNA | AK032091.1 | - | 12 - 13 | 1522 | 120 | |
AK038313.1 | - | 17 - 18 | 2301 | 120 | ||
BC027357.1 | - | 16 - 17 | 2317 | 4952415 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |